Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kcnip1 Mouse anti-Mouse, Rat, APC, Clone:G9, Abnova™

Mouse monoclonal antibody raised against full length recombinant rat Kcnip1.

Manufacturer:  Abnova Corporation MAB17452

Catalog No. 89-334-128


Add to cart

Specifications

Specifications

Kcnip1
G9
Kv channel-interacting protein 1
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Liquid
Mouse
IgG1
Store at 4°C.
Monoclonal
Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
APC
Immunocytochemistry (1:1000)
Immunofluorescence (1:1000)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000)
Western Blot (1:40)
The optimal working dilution should be determined by the end user.
Kcnip1
Kchip1
Recombinant protein corresponding to full length rat Kcnip1.
100μg
Antibody
65023
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit