Learn More
Aviva Systems Biology Corp KCNJ16 ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP35285_P050
This is a rabbit polyclonal antibody against KCNJ16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KCNJ16. Alternate names: BIR9, KIR5.1, MGC33717ApplicationWB. Species reactivity: Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-014-55690