Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNJ3 Mouse anti-Human, Clone: OTI2E4, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504210

Catalog No. 50-167-4941


Add to cart

Description

Description

KCNJ3 is a potassium channel protein controlled by G proteins. Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium. This receptor plays a crucial role in regulating the heartbeat.
Specifications

Specifications

KCNJ3
Monoclonal
0.66 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P48549
GIRK1, KGA, KIR3.1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3760
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI2E4
Unconjugated
KCNJ3
Liquid
KCNJ3
Human recombit protein fragment corresponding to amino acids 279-501 of human KCNJ3 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit