Learn More
KCNJ5 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 180631AP
Catalog No. 50-173-1965
Add to cart
Description
Description
G protein-coupled inwardly rectifying potassium channels (Kir3.1 through Kir3.4) are coupled to numerous neurotransmitter receptors in the brain and are abundantly expressed in the olfactory bulb, hippocampus, neocortex, dentate gyrus, cerebellar cortex and thalamus regions of the brain. Also known as GIRK, Kir3 potassium channels localize to the soma and dendrites as well as axons of neurons. Liberated Gbg subunits from G protein heterotrimers bind to and regulate Kir3 channel activity. Gb3- and Gb4-containing Gbg dimers bind directly to cytoplasmic domains of Kir3 proteins and increase the K+ current while Gb5-containing Gbg dimers inhibit Kir3 K+ current. Kir3 activity is also inhibited by tyrosine phosphorylation. Brain-derived neurotrophic factor activates receptor tyrosine kinase B, which then phosphorylates Kir3 tyrosine residues, effectively inactivating the Kir3 channels.
Specifications
Specifications
KCNJ5
0.15 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P48544
Cardiac inward rectifier, CIR, GIRK 4, GIRK4, Heart KATP channel, IRK 4, KATP 1, KATP1, KCNJ5, KIR3.4
Rabbit
IgG
150 μL
-20°C
Primary
3762
Western Blot
Unconjugated
KCNJ5
Liquid
KCNJ5
KCNJ5 Fusion Protein Ag12656
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human