Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP304907100UL

 View more versions of this product

Catalog No. NB093448


Add to cart

Description

Description

KCNK1 Polyclonal antibody specifically detects KCNK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

KCNK1
Unconjugated
Purified
Rabbit
IgG
100 μL
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
Immunohistochemistry, Western Blot
PBS with 50% glycerol, pH7.3.
DPK, HOHO1, Inward rectifying potassium channel protein TWIK-1, K2P1, K2p1.1, KCNO1, Potassium channel KCNO1, potassium channel subfamily K member 1, potassium channel, subfamily K, member 1, potassium inwardly-rectifying channel, subfamily K, member 1, TWIK-1, TWIK1HOHO
Recombinant fusion protein containing a sequence corresponding to amino acids 267-336 of human KCNK1 (NP_002236.1).
Affinity purified
RUO
Primary
3775
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit