Learn More
KCNK3 Mouse anti-Human, Mouse, Rat, DyLight 755, Clone: S374-48, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242202IR
Catalog No. NBP242202IR
Add to cart
Description
Description
KCNK3 Monoclonal antibody specifically detects KCNK3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
KCNK3
S374-48
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
KCNK3
Fusion protein amino acids 251-411 (cytoplasmic C-terminus) of rat Acid-sensitive potassium channel protein TASK or TASK1. Mouse: 96% identity (156/161 amino acids identical). Human: 76% identity (163/161 amino acids identical). <30% identity with TASK3.
Protein G purified
Store at 4C in the dark.
Monoclonal
Less then 30% identity with TASK3.Detects approx 50kDa. Does not cross-react with TASK3.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
Acid-sensitive potassium channel protein TASK-1, cardiac potassium channel, K2p3.1, OAT1, potassium channel subfamily K member 3, potassium channel, subfamily K, member 3, potassium inwardly-rectifying channel, subfamily K, member 3, TASK1, TASK-1, TASKacid-sensitive potassium channel protein TASK, TBAK1, TWIK-related acid-sensitive K(+) channel 1, TWIK-related acid-sensitive K+ channel, two P domain potassium channel, Two pore K(+) channel KT3.1, Two pore potassium channel KT3.1
Mouse
IgG2b
0.1 ml
Primary
3777
Human, Mouse, Rat