Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNK9 Mouse anti-Human, Clone: Y4, MilliporeSigma™

Mouse Monoclonal Antibody

$156.60 - $373.00

Specifications

Antigen KCNK9
Clone Y4
Concentration 0.5 mg/mL
Applications Flow Cytometry, Functional Assay, Inhibition Assays, Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABC1743100
View Documents
MilliporeSigma
MABC1743100UL
100 μL
Each for $373.00
Add to cart
 
MABC174325
View Documents
MilliporeSigma
MABC174325UL
25 μL
Each for $156.60
Add to cart
 
Description

Description

KCNK9 Monoclonal antibody specifically detects KCNK9 in Human samples. It is validated for Flow Cytometry, Function Assay, Inhibition, Western Blotting.
Specifications

Specifications

KCNK9
0.5 mg/mL
Unconjugated
Mouse
Human
Recombinant fragment corresponding to the 59 amino acids from the first extracellular domain (M1P1 loop) of human Potassium channel subfamily K member 9 (KCNK9).
Monoclonal
Protein G purified
Y4
Flow Cytometry, Functional Assay, Inhibition Assays, Western Blot
Purified
RUO
Purified mouse monoclonal antibody IgG1 in PBS without azide
IgG1
Primary
Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit