Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCNMB2 Monoclonal Antibody (S53-32), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA527646
Description
MaxiK channels are large conductance, voltage and calcium-sensitive potassium channels which are fundamental to the control of smooth muscle tone and neuronal excitability. MaxiK channels can be formed by 2 subunits: the pore-forming alpha subunit and the modulatory beta subunit. The protein encoded by this gene is an auxiliary beta subunit which decreases the activation time of MaxiK alpha subunit currents. Alternative splicing results in multiple transcript variants of this gene. Additional variants are discussed in the literature, but their full length nature has not been described.Specifications
KCNMB2 | |
Monoclonal | |
1 mg/ml | |
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 | |
Q811Q0, Q9CZM9, Q9Y691 | |
Kcnmb2 | |
Fusion protein amino acids 1-41 (N-terminus) and 218-235 (C-terminus) of mouse BKBeta2 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
S53-32 | |
Unconjugated | |
KCNMB2 | |
KCNMB2, Calcium-activated potassium channel subunit beta-2, BK channel subunit beta-2, BKbeta2, Calcium-activated potassium channel, subfamily M subunit beta-2, Charybdotoxin receptor subunit beta-2, K(VCA)beta-2, Maxi K channel subunit beta-2, Slo-beta-2 | |
Mouse | |
Protein G | |
RUO | |
10242, 294961, 72413 | |
-20°C | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction