Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCNMB2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA587770
Description
Immunogen sequence: TLLRSYMQSV WTEESQCTLL NASITETFNC SFSCGPDCWK LSQYPCLQVY VNLTSSGEKL LLYHTEETIK INQKCSYIPK CGKNFEESMS LVNVVMENFR KYQHFSCYSD PEGNQKSVIL TKLYSSN; Positive Samples: Mouse kidney, Mouse brain, Mouse lung, Rat brain, Rat kidney; Cellular Location: Membrane, Multi-pass membrane protein
MaxiK channels are large conductance, voltage and calcium-sensitive potassium channels which are fundamental to the control of smooth muscle tone and neuronal excitability. MaxiK channels can be formed by 2 subunits: the pore-forming alpha subunit and the modulatory beta subunit. The protein encoded by this gene is an auxiliary beta subunit which decreases the activation time of MaxiK alpha subunit currents. Alternative splicing results in multiple transcript variants of this gene. Additional variants are discussed in the literature, but their full length nature has not been described.Specifications
KCNMB2 | |
Polyclonal | |
Unconjugated | |
KCNMB2 | |
Kcnmb2 | |
Recombit fusion protein containing a sequence corresponding to amino acids 68-194 of human KCNMB2 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
2.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q811Q0, Q9CZM9, Q9Y691 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10242, 294961, 72413 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title