Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNMB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen KCNMB2
Applications Immunohistochemistry, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093520
View Documents
Novus Biologicals
NBP305145100UL
100 μg Each for $320.71
Add to cart
 
NB093521
View Documents
Novus Biologicals
NBP30514520UL
20 μg Each for $138.37
Add to cart
 
Description

Description

KCNMB2 Polyclonal antibody specifically detects KCNMB2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications

Specifications

KCNMB2
Unconjugated
Rabbit
Human, Mouse, Rat
BK channel subunit beta-2, BKbeta2, calcium-activated potassium channel subunit beta-2, Calcium-activated potassium channel, subfamily M subunit beta-2, Charybdotoxin receptor subunit beta-2, hbeta2, Hbeta3, k(VCA)beta-2, large conductance calcium-activated potassium channel beta 2 subunit, large-conductance Ca2+-activated K+ channel beta2 subunit, Maxi K channel subunit beta-2, MaxiK channel beta 2 subunit, MGC22431, potassium large conductance calcium-activated channel, subfamily M, beta member2, slo-beta-2
Recombinant fusion protein containing a sequence corresponding to amino acids 68-194 of human KCNMB2 (NP_005823.1).
Polyclonal
Affinity purified
Immunohistochemistry, Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
10242
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit