Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNMB4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP310360100UL

 View more versions of this product

Catalog No. NB125621


Add to cart

Description

Description

KCNMB4 Polyclonal antibody specifically detects KCNMB4 in Human samples. It is validated for Western Blot
Specifications

Specifications

KCNMB4
Unconjugated
PBS buffer, 2% sucrose
BK channel subunit beta-4, BKbeta4, calcium-activated potassium channel subunit beta-4, Calcium-activated potassium channel, subfamily M subunit beta-4, Charybdotoxin receptor subunit beta-4, hbeta4, K(VCA)beta-4, large conductance calcium-dependent potassium ion channel beta 4 subunit, Maxi K channel subunit beta-4, potassium large conductance calcium-activated channel, subfamily M, beta member4, slo-beta-4
The immunogen is a synthetic peptide directed towards the middle region of human KCNMB4 (NP_055320). Peptide sequence TCGADCRGTSQYPCVQVYVNNSESNSRALLHSDEHQLLTNPKCSYIPPCK
100 μg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Western Blot 1.0 ug/ml
Purified
Rabbit
Affinity purified
RUO
Primary
27345
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit