Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCNN1 Polyclonal Antibody
KCNN1 Polyclonal Antibody
Supplier: Thermo Scientific PA597206
Description
KCNN1 Polyclonal Antibody for Western Blot

Specifications
KCNN1 | |
Polyclonal | |
Unconjugated | |
Kcnn1 | |
AI854248; hSK1; KCa2.1; Kcnn1; potassium calcium-activated channel subfamily N member 1; potassium channel, calcium activated intermediate/small conductance subfamily N alpha, member 1; potassium intermediate/small conductance calcium-activated channel, subfamily N, member 1; potassium intermediate/small conductance calcium-activated channel, subfamily N, member 1-like; SK; Sk1; SKCa 1; SKCa1; small conductance Ca2+-activated K+ channel type1; small conductance calcium-activated potassium channel 1; small conductance calcium-activated potassium channel protein 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3780, 84036 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.85 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q92952, Q9EQR3 | |
Kcnn1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-90 of human KCNN1 (NP_0022392). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction