Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNQ1 Mouse anti-Human, Mouse, Rat, Clone: S37A-10, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527676

Catalog No. PIMA527676


Add to cart

Description

Description

Voltage-gated K+ channels in the plasma membrane control the repolarization and the frequency of action potentials in neurons, muscles and other excitable cells. A specific K+ channel, comprised of an alpha subunit KCNQ1 and a beta subunit KCNE1, a small protein which spans the membrane only once, is predomitly expressed in the heart and in the cochlea, and is responsible for regulating the slow, depolarization-activated potassium current. Mutations in the genes encoding for KCNQ1 and KCNE1 lead to cardiac disease because they directly impair electrical signaling, and mutations in KCNQ4 are implicated in the onset of deafness. KCNQ proteins, including KCNQ1 and KCNQ4, characteristically contain six transmembrane domains and function as tetramers. KCNQ4 forms heteromeric channels with KCNQ3 and is expressed in several tissues, including the cochlea, where it is present in outer hair cells.
Specifications

Specifications

KCNQ1
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
P51787, P97414, Q9Z0N7
ATFB1, ATFB3, FLJ26167, IKs producing slow voltage-gated potassium channel subunit alpha, IKs producing slow voltage-gated potassium channel subunit alpha KvLQT1, Jervell and Lange-Nielsen syndrome 1, JLNS1, KCNA8, KCNA9, KCNQ1, KCNQ1_HUMAN, kidney and cardiac voltage dependend K+ channel, KQT-like 1, Kv1.9, Kv7.1, KVLQT1, long (electrocardiographic) QT syndrome, Ward-Romano syndrome 1, LQT, LQT1, Potassium voltage-gated channel subfamily KQT member 1, potassium voltage-gated channel KQT-like subfamily member 1, RWS, slow delayed rectifier channel subunit, SQT2, Voltage-gated potassium channel subunit Kv7.1, WRS
Mouse
IgG1
100 μg
-20°C
Primary
16535, 3784, 84020
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
S37A-10
Unconjugated
KCNQ1
Liquid
KCNQ1
Fusion protein amino acids 2-101 of human KCNQ1
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit