Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNRG Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP18010820UL

 View more versions of this product

Catalog No. NBP18010820


Add to cart

Description

Description

KCNRG Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

KCNRG
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
KCNRG
Synthetic peptide directed towards the N terminal of human KCNRG. Peptide sequence VDRDGDLFSFILDFLRTHQLLLPTEFSDYLRLQREALFYELRSLVDLLNP.
Protein A purified
RUO
Primary
283518
Western Blot
Western Blot 1:1000
NP_775876
CLLD4, potassium channel regulatorDLTET, Protein CLLD4, putative potassium channel regulatory protein
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit