Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCNV2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548506

Catalog No. PA548506

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

Potassium channel subunit. Modulates channel activity by shifting the threshold and the half-maximal activation to more negative values.
Specifications

Specifications

KCNV2
Polyclonal
PBS with 0.09% sodium azide
Q8TDN2
Potassium voltage-gated channel subfamily V member 2, Voltage-gated potassium channel subunit Kv82, KCNV2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
169522
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
KCNV2
Liquid
KCNV2
KLH conjugated synthetic peptide between 478-507 amino acids from the C-terminal region of human KCNV2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit