Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCTD10 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548570
Description
KCTD10, a rat potassium channel tetramerisation domain-containing 10 gene is a novel member of the polymerase delta-interacting protein 1 (PDIP1) gene family. KCTD10 shares significant similarity in amino acid sequence to PDIP1 and can interact with the small subunit of DNA polymerase delta and PCNA as PDIP1 does. Like PDIP1, the expression of KCTD10 gene can be induced by TNF-alpha in NIH3T3 cells.
Specifications
KCTD10 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9H3F6 | |
KCTD10 | |
KLH conjugated synthetic peptide between 273-301 amino acids from the C-terminal region of human KCTD10. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
KCTD10 | |
BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 3, hBACURD3, BTB/POZ domain-containing protein KCTD10, Potassium channel tetramerization domain-containing protein 10, KCTD10, ULR061 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83892 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction