Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCTD12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 155231AP150UL
Description
KCTD12 gene ontology annotations related to this gene include protein homooligomerization.Specifications
KCTD12 | |
Polyclonal | |
Unconjugated | |
KCTD12 | |
C13orf2, KCTD12, KIAA1778, PFET1, PFETIN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
115207, 239217, 364458 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6WVG3, Q96CX2 | |
Kctd12 | |
KCTD12 Fusion Protein Ag5716 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction