Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCTD14 Mouse anti-Human, Clone: OTI2E9, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502526

Catalog No. 50-167-3578


Add to cart

Description

Description

KCTD14 is a protein coding gene.
Specifications

Specifications

KCTD14
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9BQ13
potassium channel tetramerization domain containing 14
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
65987
Western Blot
OTI2E9
Unconjugated
KCTD14
Liquid
KCTD14
Full length human recombit protein of human KCTD14 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit