Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCTD15 Mouse anti-Human, Mouse, Clone: 2B11, Novus Biologicals™

Mouse Monoclonal Antibody

$200.00 - $300.50

Specifications

Antigen KCTD15
Clone 2B11
Concentration 1 mg/mL
Dilution Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100
Applications ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB60232100U View Documents Novus BiologicalsSupplier Diversity Partner
NBP260232100UL
100 ul Each for $300.50
Add to cart
 
NB6023250UL View Documents Novus BiologicalsSupplier Diversity Partner
NBP26023250UL
50 ul Each for $200.00
Add to cart
 
Description

Description

KCTD15 Monoclonal antibody specifically detects KCTD15 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

KCTD15
1 mg/mL
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Purified
RUO
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
79047
Recombinant human KCTD15(1-159aa) purified from E. coli.
Monoclonal
Protein G purified
2B11
Western Blot 1:1000, ELISA 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100
Unconjugated
Mouse
Human, Mouse
BTB/POZ domain-containing protein KCTD15, MGC25497, MGC2628, potassium channel tetramerisation domain containing 15
KCTD15
IgG2b Kappa
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.

RUO

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit