Learn More
Invitrogen™ KCTD15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596984
Description
Immunogen sequence: MPHRKERPSG SSLHTHGSTG TAEGGNMSRL SLTRSPVSPL AAQGIPLPAQ LTKSNAPVHI DVGGHMYTSS LATLTKYPDS RISRLFNGTE PIVLDSLKQH YFIDRDGEIF RYVLSFLRTS KLLLPDDFKD FSLLYEEARY YQLQPMVREL ERWQQEQEQR RRSRACDCLV VRVTPDLGER IALSGEKALI EEVFPETGDV MCNSVNAGWN QDPTHVIRFP LNGYCRLNSV QDVL; Positive Samples: 293T, BT-474, Mouse skeletal muscle.
Childhood and adult obesity in the United States and to a lesser extent the rest of the world has increased dramatically over the past decade. Both environmental and genetic factors are involved in the onset and progression of weight gain. Recently, the potassium channel KCTD15 was identified as a genetic loci associated with higher than normal body mass index (BMI) in humans along with genes such as GNPDA2, MTCH2, FTO, and TMEM18. Further studies on single nucleotide polymorphisms (SNPs) in non-diabetic and diabetic patients showed that FTO was most strongly associated with obesity while MTCH2 and GNPDA2 were still significantly associated with higher than normal BMI levels. At least two isoforms of KCTD15 are known to exist.
Specifications
KCTD15 | |
Polyclonal | |
Unconjugated | |
KCTD15 | |
BC031749; BTB/POZ domain-containing protein KCTD15; Kctd15; potassium channel tetramerisation domain containing 15; potassium channel tetramerization domain containing 15; potassium channel tetramerization domain-containing protein 15 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
233107, 499129, 79047 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
3.97 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K0E1, Q96SI1 | |
KCTD15 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-234 of human KCTD15 (NP_0769812). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.