Learn More
Aviva Systems Biology Corp KCTD17 ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP35359_P050
This is a rabbit polyclonal antibody against KCTD17. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human KCTD17. Alternate names: FLJ12242ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-53917