Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCTD19 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS16930R

Specifications
KCTD19 | |
Polyclonal | |
Unconjugated | |
KCTD19 | |
4922504H04Rik; BTB/POZ domain-containing protein KCTD19; Kctd19; potassium channel tetramerisation domain containing 19; potassium channel tetramerization domain containing 19; testicular tissue protein Li 101 | |
Rabbit | |
Protein A | |
RUO | |
146212 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q17RG1 | |
KCTD19 | |
KLH conjugated synthetic peptide derived from human KCTD19. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction