Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCTD19 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | KCTD19 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP191544
|
Novus Biologicals
NBP191544 |
100 μL |
Each of 1 for $482.50
|
|
|||||
Description
KCTD19 Polyclonal specifically detects KCTD19 in Human samples. It is validated for Western Blot.Specifications
KCTD19 | |
Polyclonal | |
Rabbit | |
BTB/POZ domain-containing protein KCTD19, FLJ40162, potassium channel tetramerisation domain containing 19 | |
KCTD19 | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
146212 | |
Synthetic peptide directed towards the n terminal of human KCTD19. Peptide sequence DSLLWKEASALTSSESQRLFIDRDGSTFRHVHYYLYTSKLSFSSCAELNL. | |
Primary |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title