Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCTD2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548226

Catalog No. PA548226


Add to cart

Description

Description

Predicted to react with bovine, mouse and rat based on sequence homology.

Specifications

Specifications

KCTD2
Polyclonal
PBS with 0.09% sodium azide
Q14681
BTB/POZ domain-containing protein KCTD2, Potassium channel tetramerization domain-containing protein 2, KCTD2, KIAA0176
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23510
Western Blot
Unconjugated
KCTD2
Liquid
KCTD2
KLH conjugated synthetic peptide between 234-262 amino acids from the C-terminal region of human KCTD2.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit