Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCTD20 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549277

Catalog No. PA549277

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Promotes the phosphorylation of AKT family members.
Specifications

Specifications

KCTD20
Polyclonal
PBS with 0.09% sodium azide
Q8CDD8
BTB/POZ domain-containing protein KCTD20, Potassium channel tetramerization domain containing 20, KCTD20, C6orf69
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
66989
Western Blot
Unconjugated
Kctd20
Liquid
KCTD20
KLH conjugated synthetic peptide between 313-341 amino acids from the C-terminal region of human KCTD20.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit