Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KCTD21 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | KCTD21 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179549
|
Novus Biologicals
NBP179549 |
100 μL |
Each of 1 for $436.00
|
|
Description
KCTD21 Polyclonal specifically detects KCTD21 in Human samples. It is validated for Western Blot.Specifications
KCTD21 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
BTB/POZ domain-containing protein KCTD21, KCASH2, potassium channel tetramerisation domain containing 21 | |
KCTD21 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
NP_001025030 | |
283219 | |
Synthetic peptide directed towards the middle region of human KCTD21The immunogen for this antibody is KCTD21. Peptide sequence VFNANIFSTSCLFLKLLGSKLFYCSNGNLSSITSHLQDPNHLTLDWVANV. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title