Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCTD4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$436.00

Specifications

Antigen KCTD4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP180106
View Documents
Novus Biologicals
NBP180106
100 μL
Each of 1 for $436.00
Add to cart
 
Description

Description

KCTD4 Polyclonal specifically detects KCTD4 in Human samples. It is validated for Western Blot.
Specifications

Specifications

KCTD4
Polyclonal
Rabbit
NP_940686
386618
Synthetic peptide directed towards the middle region of human KCTD4. Peptide sequence EITDNHDRSQGLRIFCNAPDFISKIKSRIVLVSKSRLDGFPEEFSISSNI.
Primary
Store at -20C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
RUO
bA321C24.3, BTB/POZ domain-containing protein KCTD4, potassium channel tetramerisation domain containing 4
KCTD4
IgG
Affinity Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit