Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KCTD5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100587
Description
Antibody detects endogenous levels of total KCTD5.
KCTD5 is involved in interaction with CUL3 suggests that it may act as a substrate adapter in some E3 ligase complex. Does not affect the function of Kv channel Kv2.1/KCNB1, Kv1.2/KCNA2, Kv4.2/KCND2 and Kv3.4/KCNC4.
Specifications
KCTD5 | |
Polyclonal | |
Unconjugated | |
KCTD5 | |
2610030N08Rik; BTB/POZ domain-containing protein KCTD5; KCTD5; mKIAA0176; potassium channel tetramerisation domain containing 5; potassium channel tetramerization domain containing 5 | |
Rabbit | |
Affinity chromatography | |
RUO | |
287109, 54442, 69259 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
B5DEL1, Q8VC57, Q9NXV2 | |
KCTD5 | |
A synthesized peptide derived from human KCTD5(Accession Q9NXV2), corresponding to amino acid residues E7-T57. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction