Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCTD8 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548245

Catalog No. PA548245

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Auxiliary subunit of GABA-B receptors that determine the pharmacology and kinetics of the receptor response. Increases agonist potency and markedly alter the G-protein signaling of the receptors by accelerating onset and promoting desensitization.
Specifications

Specifications

KCTD8
Polyclonal
PBS with 0.09% sodium azide
Q6ZWB6, Q50H33
BTB/POZ domain-containing protein KCTD8, KCTD8
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
243043, 386617
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
KCTD8
Liquid
KCTD8
KLH conjugated synthetic peptide between 434-462 amino acids from the C-terminal region of human KCTD8.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit