Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KCTD9 Mouse anti-Human, Clone: OTI2G8, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526902

Catalog No. PIMA526902


Add to cart

Description

Description

Substrate-specific adapter of a BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex, which mediates the ubiquitination of target proteins, leading to their degradation by the proteasome.
Specifications

Specifications

KCTD9
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q7L273
BTBD27; BTB/POZ domain-containing protein KCTD9; potassium channel tetramerisation domain containing 9
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54793
Immunohistochemistry (Paraffin), Western Blot
OTI2G8
Unconjugated
KCTD9
Liquid
KCTD9
Human recombit protein fragment corresponding to amino acids 1-223 of KCTD9 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit