Learn More
KD Scientific Allegro™ I Peristaltic Pump System
Features single channel pump head
Manufacturer:  KD Scientific 785501
Catalog No. 14-831-400
Description
Description
  • Allows maintenance free, continuous flow pumping of various liquids
  • Fluid contacts only inside surface of tubing negating concern for valves, O-rings or seals that might be incompatible with fluid being pumped
  • Clearly displays flow rate and total dispensed volume
  • User configurable multi-step operation
  • Dispenses in forward or reverse direction
  • Adjustable flow rate while pump is running
  • Stackable sealed remote pump drive with IP 32 rating
  • Selectable tubing table
  • Automatic tubing break-in and calibration procedures
  • External connectivity with USB interface and I/O port
  • Flow rates up to 1500mL/min.
  • Accepts tubing ID sizes 0.8mm (0.03 in.) to 8.0mm (0.32 in.)
  • Compatible with Masterflex™ L/S 13, L/S 14, L/S 16, L/S 25, L/S 17 and L/S 18 tubing
Specifications
Specifications
1500 mL/min. max.