Learn More
KD Scientific KDS310 Infusion Withdrawal Pump
Features small size, remote control and a rugged mounting arm
Manufacturer:  KD Scientific 780311
Catalog No. 14-831-16
Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Mini size pump supports a 0.5 to 250μL microliter syringe (not included)
  • Simple menu-driven setup with bright display and keypad allows selection of syringe size, dispense volume and flow rate, infusion or withdrawal
  • Dispense volume is displayed and has automatic shutoff at completion
  • Easy syringe loading with pusher block fast forward/fast reverse feature for rapid adjustments
  • Last settings stored in memory
Specifications
Specifications
110 V
0.9 kg
Infusion Withdrawal Pump
±<1%
1.7 in.
6.5 in.
1.18 in.
0.5 to 250 μL
2 lb.
Ideal for use with micromanipulator, stereotaxic and other clamping devices
1
4.5 cm
16.5 cm
SDS
Documents
Documents
Product Certifications
Accessories