Learn More
KD Scientific Legacy KDS 270 Continuous Cycle Pump Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  KD Scientific 789270
 View more versions of this product
Catalog No. 14-931-30
Add to cart
Description
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
220 V
9.5 lb.
220 VAC 50/60 Hz
Backlit LCD
CE
4 kg
Electrospinning, microfluids, electrospray, mass spec, continuous flow, organic synthesis, drug infusion, Maldi-TOF, emulsifier
±<1%
SDS
Documents
Product Certifications
Accessories