Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KDEL Mouse anti-Chemical, Clone: 1F2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527579

Catalog No. PIMA527579


Add to cart

Description

Description

The sequence Lys-Asp-Glu-Leu (KDEL) or a closely related sequence, is present at the carboxy-terminus of soluble endoplasmic reticulum (ER) resident proteins and some membrane proteins. 78 and 94 kDa glucose regulated proteins, GRP78 and GRP94 respectively, and protein disulfide isomerase (PDI) all share the C-terminal KDEL sequence. The presence of carboxy-terminal KDEL appears to be necessary for ER retention and appears to be sufficient to reduce the secretion of proteins from the ER. This retention is reported to be mediated by a KDEL receptor.
Specifications

Specifications

KDEL
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
KDEL, Lys Asp Glu Leu, Lys-Asp-Glu-Leu (KDEL), Lys-Asp-Glu-Leu, Lysine-aspartic acid-glutamate-leucine
KDEL peptide (C-AVKDEL -COOH) conjugated at the N-terminal to Bovine Serum Albumin (BSA)
Protein G
RUO
Antibody
Monoclonal
Western Blot
1F2
Unconjugated
Liquid
Mouse
IgG1
100 μg
-20°C
Primary
Chemical
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit