Learn More
KDEL Mouse anti-Human, Mouse, Rat, DyLight 350, Clone: 2D6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP261927UV
Catalog No. NBP261927UV
Add to cart
Description
Description
KDEL Monoclonal antibody specifically detects KDEL in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
Specifications
KDEL
2D6
Western Blot
Purified
KDELC1
KDEL peptide (C-AVKDEL -COOH) conjugated at the N-terminal to Bovine Serum Albumin (BSA)
Protein G purified
Cellular Markers, ER Markers, Membrane Trafficking and Chaperones
Primary
79070
Human, Mouse, Rat
Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
Endoplasmic reticulum resident protein 58, EP58KDEL1, ER protein 58, ERp58, KDEL (Lys-Asp-Glu-Leu) containing 1, KDEL motif-containing 1, KDEL motif-containing protein 1, MGC5302
Mouse
IgG1
100 ul
Store at 4C in the dark.
Monoclonal
Detects KDEL tagged protein.