Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KDEL Antibody (1F2), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261924V
Description
KDEL Monoclonal specifically detects KDEL in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
KDEL | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
KDELC1 | |
KDEL peptide (C-AVKDEL -COOH) conjugated at the N-terminal to Bovine Serum Albumin (BSA) | |
100 μL | |
Primary | |
Detects KDEL tagged proteins. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot | |
1F2 | |
Western Blot | |
Endoplasmic reticulum resident protein 58, EP58KDEL1, ER protein 58, ERp58, KDEL (Lys-Asp-Glu-Leu) containing 1, KDEL motif-containing 1, KDEL motif-containing protein 1, MGC5302 | |
Mouse | |
Protein G purified | |
Cellular Markers, ER Markers, Membrane Trafficking and Chaperones | |
79070 | |
Human, Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction