Learn More
KDEL Mouse anti-Human, Mouse, Rat, DyLight 488, Clone: 2C1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP261926G
Catalog No. NBP261926G
Add to cart
Description
Description
KDEL Monoclonal antibody specifically detects KDEL in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
Specifications
KDEL
2C1
Western Blot, Immunohistochemistry
50mM Sodium Borate with 0.05% Sodium Azide
79070
Mouse
IgG1
Primary
100 ul
Store at 4C in the dark.
Detects KDEL tagged proteins.
Western Blot,Immunohistochemistry
DyLight 488
Purified
Endoplasmic reticulum resident protein 58, EP58KDEL1, ER protein 58, ERp58, KDEL (Lys-Asp-Glu-Leu) containing 1, KDEL motif-containing 1, KDEL motif-containing protein 1, MGC5302
KDELC1
KDEL peptide (C-AVKDEL -COOH) conjugated at the N-terminal to Bovine Serum Albumin (BSA)
Monoclonal
Protein G purified
Cellular Markers, ER Markers, Membrane Trafficking and Chaperones
Human,Mouse,Rat