Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KDEL Mouse anti-Human, Mouse, Rat, DyLight 680, Clone: 2D6, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP261927FR
Description
KDEL Monoclonal antibody specifically detects KDEL in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
KDEL | |
2D6 | |
Western Blot | |
Purified | |
KDELC1 | |
KDEL peptide (C-AVKDEL -COOH) conjugated at the N-terminal to Bovine Serum Albumin (BSA) | |
Protein G purified | |
Cellular Markers, ER Markers, Membrane Trafficking and Chaperones | |
Primary | |
79070 | |
Human, Mouse, Rat |
Western Blot | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Endoplasmic reticulum resident protein 58, EP58KDEL1, ER protein 58, ERp58, KDEL (Lys-Asp-Glu-Leu) containing 1, KDEL motif-containing 1, KDEL motif-containing protein 1, MGC5302 | |
Mouse | |
IgG1 | |
100 ul | |
Store at 4C in the dark. | |
Monoclonal | |
Detects KDEL tagged protein. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title