Learn More
KDELC2 Mouse anti-Human, Clone: OTI5G2, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809568
Catalog No. 50-166-9591
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
KDELC2 is a protein coding gene.
Specifications
Specifications
KDELC2
Monoclonal
Unconjugated
KDELC2
Lyophilized
KDELC2
Full length human recombit protein of human KDELC2 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI5G2
PBS with 8% trehalose and no preservative; pH 7.3
Q7Z4H8
KDEL motif containing 2
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
143888