Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KDM4A Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 249431AP

Catalog No. 50-173-4633


Add to cart

Description

Description

This gene is a member of the Jumonji domain 2 family and encodes a protein containing a JmjN domain, a JmjC domain, a JD2H domain, two TUDOR domains, and two PHD-type zinc fingers. This nuclear protein functions as a trimethylation-specific demethylase, converting specific trimethylated histone residues to the dimethylated form, and as a transcriptional repressor.
Specifications

Specifications

KDM4A
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O75164, Q8BW72
JHDM3A, JMJD2, JMJD2A, KDM4A, KIAA0677, lysine (K) demethylase 4A, Lysine specific demethylase 4A
Rabbit
IgG
150 μL
-20°C
Primary
230674, 9682
Western Blot
Unconjugated
KDM4A
Liquid
KDM4A
KDM4A Fusion Protein Ag20747
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit