Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KDM4D Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 225911AP150UL
Description
JMJD2D belongs to JMJD2 family that is consist of four cancer-associated genes.Specifications
KDM4D | |
Polyclonal | |
Unconjugated | |
KDM4D | |
JHDM3D, JMJD2D, KDM4D, lysine (K) demethylase 4D, Lysine specific demethylase 4D | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55693 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6B0I6 | |
KDM4D | |
KDM4D Fusion Protein Ag18436 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction