Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KDM6A Mouse anti-Human, Biotin, Clone: 1612, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243665B

Catalog No. NBP243665B


Add to cart

Description

Description

KDM6A Monoclonal antibody specifically detects KDM6A in Human samples. It is validated for Western Blot.
Specifications

Specifications

KDM6A
1612
Western Blot
Purified
KDM6A
Recombinant protein encompassing a sequence within the center region of human KDM6A. The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Western Blot
Biotin
PBS with 0.05% Sodium Azide
bA386N14.2, bA386N14.2 (ubiquitously transcribed X chromosome tetratricopeptide repeatprotein (UTX)), DKFZp686A03225, EC 1.14.11, EC 1.14.11.-, Histone demethylase UTX, lysine (K)-specific demethylase 6A, MGC141941, ubiquitously transcribed tetratricopeptide repeat protein X-linked, ubiquitously transcribed tetratricopeptide repeat, X chromosome, ubiquitously transcribed TPR protein on the X chromosome, ubiquitously transcribed X chromosome tetratricopeptide repeat protein, ubiquitously-transcribed TPR gene on the X chromosome, Ubiquitously-transcribed TPR protein on the X chromosome, Ubiquitously-transcribed X chromosome tetratricopeptide repeat protein, UTXlysine-specific demethylase 6A
Mouse
IgG1
0.1 ml
Primary
7403
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit