Learn More
KDR Mouse anti-Human, Clone: OTI2A1, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF802443
Catalog No. 50-166-6016
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Vascular endothelial growth factor (VEGF) is a major growth factor for endothelial cells. This gene encodes one of the two receptors of the VEGF. This receptor, known as kinase insert domain receptor, is a type III receptor tyrosine kinase. It functions as the main mediator of VEGF-induced endothelial proliferation, survival, migration, tubular morphogenesis and sprouting. The signalling and trafficking of this receptor are regulated by multiple factors, including Rab GTPase, P2Y purine nucleotide receptor, integrin alphaVbeta3, T-cell protein tyrosine phosphatase, etc.. Mutations of this gene are implicated in infantile capillary hemangiomas.
Specifications
Specifications
KDR
Monoclonal
Unconjugated
KDR
Lyophilized
KDR
Human recombit protein fragment corresponding to amino acids 786-1356 of human KDR produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI2A1
PBS with 8% trehalose and no preservative; pH 7.3
P35968
CD309, FLK1, VEGFR, VEGFR2
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
3791