Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KDR (VEGFR2) Recombinant Human Protein Non-distribution product as customer accommodation.

KDR (VEGFR2) Recombinant Human Protein

$433.00

Specifications

Accession Number NP_002244
Assays KDR (VEGFR2) was pre-diluted in enzyme dilution buffer and assayed in 50mM Tris (pH 7.5), 10mM MgCl2, 0.25mM EGTA, 0.1mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
Common Name KDR (VEGFR2) Recombinant Human Protein
Concentration 0.43mg/mL
Expression System Baculovirus
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PV3660 Non-distribution product as customer accommodation. View Documents LIFE TECHNOLOGIES
PV3660
10μg Each for $433.00
Add to cart
 
Description

Description

KDR (VEGFR2) is a type III receptor tyrosine kinase. It binds vascular endothelial growth factor with high affinity in vitro, and is expressed early in development by endothelial cell precursors.
Specifications

Specifications

NP_002244
KDR (VEGFR2) Recombinant Human Protein
Baculovirus
VEGF receptors
KDR (VEGFR2) Recombinant Human Protein
KDR (VEGFR2) Recombinant Human Protein
80% by SDS-PAGE
892nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL
Kinase assay
His Tag
KDR (VEGFR2) was pre-diluted in enzyme dilution buffer and assayed in 50mM Tris (pH 7.5), 10mM MgCl2, 0.25mM EGTA, 0.1mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C
0.43mg/mL
KDR (VEGFR2), VEGFR
68.4kDa
1 Each
Amino acids 789-1356
For research use only. Not for use in diagnostic procedures
–80°C
Human

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit