Learn More
Keap1 Mouse anti-Human, Alexa Fluor 405, Clone: 7G4B10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237431AF405
Catalog No. NBP237431N
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Keap1
7G4B10
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Purified
KEAP1
Purified recombinant fragment of human KEAP1 (AA: 380-624 ) expressed in E. Coli.
Protein A or G purified
Stem Cell Markers
Primary
9817
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
INRF2, INrf2MGC9454, kelch-like ECH-associated protein 1, Kelch-like protein 19, KIAA0132MGC4407, KLHL19Cytosolic inhibitor of Nrf2, MGC10630, MGC1114, MGC20887
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human