Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keap1 Mouse anti-Human, FITC, Clone: 7G4B10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237431F

Catalog No. NBP237431F

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Keap1
7G4B10
Flow Cytometry, ELISA, Immunohistochemistry
Purified
KEAP1
Purified recombinant fragment of human KEAP1 (AA: 380-624 ) expressed in E. Coli.
Protein A or G purified
Stem Cell Markers
Primary
9817
Flow Cytometry, ELISA, Immunohistochemistry
FITC
PBS with 0.05% Sodium Azide
INRF2, INrf2MGC9454, kelch-like ECH-associated protein 1, Kelch-like protein 19, KIAA0132MGC4407, KLHL19Cytosolic inhibitor of Nrf2, MGC10630, MGC1114, MGC20887
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit