Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Keap1 Antibody (OTI1B4), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271496AF647
Description
Keap1 Monoclonal specifically detects Keap1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
Keap1 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
KEAP1 | |
Full length recombinant protein of human Keap1 (NP_987096) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Monkey | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI1B4 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
INRF2, INrf2MGC9454, kelch-like ECH-associated protein 1, Kelch-like protein 19, KIAA0132MGC4407, KLHL19Cytosolic inhibitor of Nrf2, MGC10630, MGC1114, MGC20887 | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
9817.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction