Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KEAP1 Mouse anti-Human, Mouse, Clone: OTI1B4, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502059S

Catalog No. 50-167-3146


Add to cart

Description

Description

This gene encodes a protein containing KELCH-1 like domains, as well as a BTB/POZ domain. Kelch-like ECH-associated protein 1 interacts with NF-E2-related factor 2 in a redox-sensitive manner and the dissociation of the proteins in the cytoplasm is followed by transportation of NF-E2-related factor 2 to the nucleus. This interaction results in the expression of the catalytic subunit of gamma-glutamylcysteine synthetase. Two alternatively spliced transcript variants encoding the same isoform have been found for this gene.
Specifications

Specifications

KEAP1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q14145, Q9Z2X8
INrf2, KLHL19
Mouse
IgG1
30 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
50868, 9817
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI1B4
Unconjugated
KEAP1
Liquid
Keap1
Full length human recombit protein of human KEAP1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit