Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keap1 Mouse anti-Human, Mouse, Janelia Fluor™ 549, Clone: OTI1B4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271496JF549

Catalog No. N271496J549


Add to cart

Description

Description

Keap1 Monoclonal antibody specifically detects Keap1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications

Specifications

Keap1
OTI1B4
Flow Cytometry, Immunohistochemistry
Affinity Purified, Conjugated
KEAP1
Full length recombinant protein of human Keap1 (NP_987096) produced in HEK293T cells.
Immunogen affinity purified
Stem Cell Markers
Primary
9817.0
Flow Cytometry, Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
INRF2, INrf2MGC9454, kelch-like ECH-associated protein 1, Kelch-like protein 19, KIAA0132MGC4407, KLHL19Cytosolic inhibitor of Nrf2, MGC10630, MGC1114, MGC20887
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit