Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ KEAP1 Recombinant Human Protein, N-His.GST.AVI Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human KEAP1 (Gln 2-Cys 624) fused to N-terminal polyhistidine-tagged GST tag at N-terminus linked by AVI tag

Manufacturer:  Sino Biological™ 11981H29B25

 View more versions of this product

Catalog No. 11-981-H29B25


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Expressed from DNA sequence encoding human KEAP1 (Gln 2-Cys 624) fused to N-terminal polyhistidine-tagged GST tag at N-terminus linked by AVI tag

  • Formulation: 20mM Tris, 500mM NaCl, 10% glycerol, pH 7.4.
Description & Specifications

Specifications

Analysis of protein stucture and used in cell biology research applications
9817
≥85% by SDS-PAGE
Human
<1 EU/μg
KEAP1
Other proteins
KEAP1 Recombinant Human Protein
Lyophilized
99.2kd
5 x 5μg
Full Length, Recombinant
Kelch-like ECH-Associated Protein 1
Assay Standard
Insect
N-His, GST and AVI Tags
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit